missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Salicylate, TRC

Product Code. 30386961
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386961 5 g
30371794 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386961 Supplier Toronto Research Chemicals Supplier No. TRCE9187835G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym 2-Hydroxybenzoic acid ethyl ester, Salicylic acid, ethyl ester (6CI,7CI,8CI), 2-Ethoxycarbonylphenol, 2-Hydroxybenzoic acid ethyl ester, Ethyl 2-hydroxybenzoate, Ethyl o-hydroxybenzoate, Ethyl salicylate, Mesotol, NSC 8209, Sal Ethyl, Salicylic ether, Salotan, o-(Ethoxycarbonyl)phenol, Salicylic acid-ethyl ester
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)c1ccccc1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3
Formula Weight 166.063
Chemical Name or Material Salicylic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.