missing translation for 'onlineSavingsMsg'
Learn More

Ethyl stearate, 97%

Product Code. 10378150 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10378150

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-61-5 | C20H40O2 | 312.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-61-5
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.52
MDL Number MFCD00009006
InChI Key MVLVMROFTAUDAG-UHFFFAOYSA-N
Synonym ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate
PubChem CID 8122
ChEBI CHEBI:84936
IUPAC Name ethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 30°C to 39°C
Color White
Boiling Point 213°C to 215°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)16CO2C2H5
Quantity 25 g
Beilstein 02, 379
Merck Index 15, 8930
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,acetone
Formula Weight 312.52
Percent Purity 97%
Physical Form Crystalline Mass
Chemical Name or Material Ethyl stearate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.