missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-acetate, 98%

Product Code. 11428587
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11428587

Brand: Thermo Scientific Alfa Aesar A19694.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37784-63-7 | C8H10O2S | 170.23 g/mol

Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37784-63-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00005472
InChI Key FZBNQIWPYCUPAP-UHFFFAOYSA-N
Synonym ethyl thiophene-3-acetate, ethyl 3-thienylacetate, ethyl 3-thiopheneacetate, ethyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, ethyl ester, ethyl 3-thienylacetate #, acmc-1afw1, ethyl 2-3-thienyl acetate, bidd:gt0101, ethyl 2-thiophen-3-ylethanoate
PubChem CID 520865
SMILES CCOC(=O)CC1=CSC=C1

Specifications

Density 1.137
Boiling Point 110°C to 114°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.51
Quantity 5 g
Beilstein 122555
IUPAC Name ethyl 2-(thiophen-3-yl)acetate
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.