missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-bromocinnamate, 98%

Product Code. 11481830
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481830 5 g 5g
11491830 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481830 Supplier Thermo Scientific Alfa Aesar Supplier No. H27544.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24393-53-1 | C11H11BrO2 | 255.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24393-53-1
Molecular Formula C11H11BrO2
Molecular Weight (g/mol) 255.111
MDL Number MFCD00012226
InChI Key YOOKYIPLSLPRTC-VMPITWQZSA-N
Synonym ethyl trans-4-bromocinnamate, e-ethyl 3-4-bromophenyl acrylate, ethyl 3-4-bromophenyl acrylate, 2-propenoic acid, 3-4-bromophenyl-, ethyl ester, ethyl 2e-3-4-bromophenyl prop-2-enoate, 2-propenoic acid, 3-4-bromophenyl-, ethyl ester, 2e, ethyl p-bromocinnamate, ethyl e-4-bromocinnamate, ethyl-trans-4-bromocinnamate, 4-bromocinnamic acid ethyl ester
PubChem CID 688115
IUPAC Name ethyl (E)-3-(4-bromophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)Br

Specifications

Density 1.359
Boiling Point 180°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 5 g
Formula Weight 255.11
Percent Purity 98%
Chemical Name or Material Ethyl trans-4-bromocinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.