Learn More
Ethyl trans-4-oxo-2-butenoate, 96%
Beschreibung
Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.
Solubility
Slightly soluble in water(2.6 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Spezifikation
Spezifikation
| Melting Point | -9°C |
| Density | 1.063 |
| Boiling Point | 184°C to 188°C |
| Flash Point | 68°C (154°F) |
| Refractive Index | 1.455 |
| Quantity | 25 g |
| UN Number | UN2922 |
| Beilstein | 8762336 |
| Solubility Information | Slightly soluble in water(2.6g/L at 25°C). |
| Formula Weight | 128.13 |
| Mehr anzeigen |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.