missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-oxo-2-butenoate, 96%

Product Code. 11355015
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11355015

Brand: Thermo Scientific Alfa Aesar L18491.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2960-66-9 | C6H8O3 | 128.127 g/mol

Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trans-4-oxo-2-butenoate is an intermediate used to synthesize pharmaceutically active molecules. It is also used in the asymmetric catalytic alkynylation of acetaldehyde, producing attractive synthons with broad applications, such as (+)-Tetrahydropyrenophorol.

Solubility
Slightly soluble in water(2.6 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2960-66-9
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD00236170
InChI Key SDGAEBKMHIPSAC-ONEGZZNKSA-N
Synonym ethyl trans-4-oxo-2-butenoate, ethyl 4-oxobut-2-enoate, e-ethyl 4-oxobut-2-enoate, ethyl e-4-oxobut-2-enoate, ethyl 2e-4-oxobut-2-enoate, e-4-oxo-but-2-enoic acid ethyl ester, 2-butenoic acid, 4-oxo-, ethyl ester, 2e, ethyl fumaraldehydate, ethyl 3-formylacrylate, ethyl-e-4-oxobutenoate
PubChem CID 7019541
IUPAC Name ethyl (E)-4-oxobut-2-enoate
SMILES CCOC(=O)C=CC=O

Specifications

Melting Point -9°C
Density 1.063
Boiling Point 184°C to 188°C
Flash Point 68°C (154°F)
Refractive Index 1.455
Quantity 100 g
UN Number UN2922
Beilstein 8762336
Solubility Information Slightly soluble in water(2.6g/L at 25°C).
Formula Weight 128.13
Percent Purity 96%
Chemical Name or Material Ethyl trans-4-oxo-2-butenoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.