missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoroacetate, TRC

Product Code. 30136604
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136604 50 g 50g
30136158 100 g 100g
30010849 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136604 Supplier Toronto Research Chemicals Supplier No. TRCE93045050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.08
Synonym 2,2,2-Trifluoroacetic Acid Ethyl Ester, Ethyl Perfluoroacetate, Ethyl Trifluoroacetate, Ethyl alpha,alpha,alpha-Trifluoroacetate, NSC 220215, Trifluoroacetic Acid Ethyl Ester;
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Quantity 50 g
InChI Formula InChI=1S/C4H5F3O2/c1-2-9-3(8)4(5,6)7/h2H2,1H3
Formula Weight 142.0242
Chemical Name or Material Ethyl Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.