Learn More
Ethyl trifluoropyruvate, 97%
Description
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl trifluoropyruvate acts as a building block in synthetic chemistry for the synthesis of fluorine containing compounds. It is also used as a reagent for the preparation of potential biologically active compounds like highly enantioselective organocatalytic hydroxyalkylation of indoles.
Solubility
Miscible with dichloromethane.
Notes
Incompatible with strong oxidizing agents, reducing agents, acids and alkalis (bases). Store in a cool place.
Spécification
Spécification
| Density | 1.4 |
| Boiling Point | 95°C to 98°C |
| Flash Point | 27°C (80°F) |
| Refractive Index | 1.3405 |
| Quantity | 25 g |
| UN Number | UN3272 |
| Beilstein | 2087388 |
| Solubility Information | Miscible with dichloromethane. |
| Formula Weight | 170.09 |
| Percent Purity | 97% |
| Afficher plus |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.