missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vanillin, TRC

Product Code. 30136640
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136640 5 g 5g
30133470 100 g 100g
30005172 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136640 Supplier Toronto Research Chemicals Supplier No. TRCE9321005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-32-4
Molecular Formula C9 H10 O3
Molecular Weight (g/mol) 166.17
Synonym Ethylvanillin, Benzaldehyde, 3-ethoxy-4-hydroxy-, 3-Ethoxy-4-hydroxybenzaldehyde, 2-Ethoxy-4-formylphenol, 3-Ethylvanillin, 4-Hydroxy-3-ethoxybenzaldehyde, Arovanillon, Bourbonal, Ethavan, Ethovan, Ethylprotal, Ethyl vanillin, Ethylvanillin, NSC 1803, NSC 67240, Protocatechuic aldehyde ethyl ether, Quantrovanil, Rhodiarome, Vanillal, Vanirom
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
SMILES CCOc1cc(C=O)ccc1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3
Formula Weight 166.063
Percent Purity >95%
Chemical Name or Material Ethylvanillin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.