missing translation for 'onlineSavingsMsg'
Learn More

Ethyl vinyl ether, 99%, stabilized

Product Code. 10523884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10523884 250 mL Glass bottle 250mL
10734421 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10523884 Supplier Thermo Scientific Acros Supplier No. 119082500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-92-2 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-92-2 , 1310-58-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00009248
InChI Key FJKIXWOMBXYWOQ-UHFFFAOYSA-N
Synonym ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
PubChem CID 8023
IUPAC Name ethenoxyethane
SMILES CCOC=C

Specifications

Melting Point -116.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 36.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH=CH2
Refractive Index 1.3760 to 1.3780
Quantity 250 mL
Beilstein 01,433
Fieser 01,386; 02,198; 04,234; 10,184; 11,235
Specific Gravity 0.75
Solubility Information Solubility in water: 7.8g/L (25°C). Other solubilities: soluble in most organic solvents
Stabilizer 0.1% KOH
Viscosity 0.28 mPa.s (5°C)
Formula Weight 72.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl vinyl ether, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl vinyl ether, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl vinyl ether, stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.