missing translation for 'onlineSavingsMsg'
Learn More

Ethylamine, 2M solution in THF, AcroSeal™

Product Code. 10136263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
Product Code. Quantity unitSize
10136263 100 mL 100mL
10390514 800 mL 800mL
2 options
This item is not returnable. View return policy

Product Code. 10136263

Brand: Thermo Scientific Acros 370251000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-04-7 | C2H7N | 45.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-04-7
InChI Key QUSNBJAOOMFDIB-UHFFFAOYSA-N
IUPAC Name ethanamine
SMILES CCN

Specifications

Density 0.8610 g/mL
Flash Point -34°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C2H7N
Linear Formula CH3CH2NH2
Quantity 100 mL
Merck Index 15, 3817
Specific Gravity 0.861
Solubility Information Solubility aq. soln.: miscible.
Molecular Weight (g/mol) 45.09
Formula Weight 45.08
Chemical Name or Material Ethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylamine, 2.0M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.