missing translation for 'onlineSavingsMsg'
Learn More

Ethylamine, Pure, Solution In Water 70% (w/w), Fisher Chemical

Product Code. 10637602 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10637602 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10637602 Supplier Fisher Chemical Supplier No. E/1000/08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-04-7
Melting Point -81.1°C
Color Colorless
pH 14
Boiling Point 39°C
Packaging Amber glass bottle
Molecular Formula C2H7N
MDL Number 8160
Quantity 500 mL
Synonym ethylamine, monoethylamine, aminoethane, 1-aminoethane, ethyl amine, aethylamine, etilamina, ethylamine solution, ethylamin, etyloamina
InChI Key QUSNBJAOOMFDIB-UHFFFAOYSA-N
SMILES CCN
IUPAC Name ethanamine
Molecular Weight (g/mol) 45.085
PubChem CID 6341
Vapor Density 1.56
ChEBI CHEBI:15862
Formula Weight 45.07g/mol
Vapor Pressure 423 hPa at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.