missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, 99%, Reagent Grade, Honeywell™

Product Code. 15637840 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15637840 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15637840 Supplier Honeywell Research Chemicals Supplier No. E125082.5L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-41-4
Melting Point -95°C
Color Colorless
Density 0.867g/mL at 25°C
Boiling Point 135°C to 137°C
Flash Point 15°C (59°F)
Odor Characteristic
Packaging Glass bottle
Molecular Formula C8H10
Linear Formula C6H5C2H5
MDL Number MFCD00011647
Quantity 2.5 L
UN Number UN1175
Beilstein 1901871
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
SMILES CCC1=CC=CC=C1
IUPAC Name ethylbenzene
Molecular Weight (g/mol) 106.168
PubChem CID 7500
ChEBI CHEBI:16101
Formula Weight 106.17g/mol
Percent Purity 0.99
Chemical Name or Material Ethylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.