missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, ≥99 %

Product Code. 11357635
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357635 2500 mL 2.5 L
11337635 100 mL 100mL
11347635 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357635 Supplier Thermo Scientific Alfa Aesar Supplier No. L05908.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-41-4 | C8H10 | 106.168 g/mol

Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylbenzene is used as a solvent for rubber adhesives, varnishes, resins, paints and coatings. It acts as an intermediate in the preparation of styrene. It is also used as a fuel additive. It is added to gasoline as an anti-knock agent, thereby reducing engine knocking and increase the octane rating. In addition to this, it is used in natural gas recovery process.

Solubility
Miscible with alcohol, ethylalcohol, benzene, ethyl ether, carbon tetrachloride, organic solvents, sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Melting Point -95°C
Density 0.867
Boiling Point 135°C to 136°C
Flash Point 15°C (59°F)
Odor Petrol-like
Refractive Index 1.495
Quantity 2500 mL
UN Number UN1175
Beilstein 1901871
Merck Index 14,3765
Solubility Information Miscible with alcohol,ethylalcohol,benzene,ethyl ether,carbon tetrachloride,organic solvents,sulfur dioxide and ether. Slightly miscible with chloroform. Immiscible with water and ammonia.
Formula Weight 106.17
Percent Purity 99%
Chemical Name or Material Ethylbenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylbenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1A
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Ethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.