missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, TRC

Product Code. 17970435
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970435 Supplier Toronto Research Chemicals Supplier No. TRCE8997451G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-41-4
Molecular Formula C8 H10
Quantity 1 g
Synonym Benzene, ethyl-, Ethylbenzene, 1-Ethylbenzene, EB, Ethylbenzol, NSC 406903, Phenylethane, α-Methyltoluene
InChI Formula InChI=1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3
SMILES CCc1ccccc1
IUPAC Name ethylbenzene
Molecular Weight (g/mol) 106.16
Formula Weight 106.0783
Chemical Name or Material Ethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.