missing translation for 'onlineSavingsMsg'
Learn More

Ethylboronic acid, 97%

Product Code. 11354336
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354336 1 g 1g
11364336 5 g 5g
11374336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354336 Supplier Thermo Scientific Alfa Aesar Supplier No. L19959.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4433-63-0 | C2H7BO2 | 73.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4433-63-0
Molecular Formula C2H7BO2
Molecular Weight (g/mol) 73.89
MDL Number MFCD01074536
InChI Key PAVZHTXVORCEHP-UHFFFAOYSA-N
Synonym boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2
PubChem CID 521157
IUPAC Name ethylboronic acid
SMILES CCB(O)O

Specifications

Melting Point 84°C to 88°C
Odor Odorless
Linear Formula CH3CH2B(OH)2
Quantity 1 g
Beilstein 1731546
Sensitivity Air sensitive
Formula Weight 73.89
Percent Purity 98%
Chemical Name or Material Ethylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.