missing translation for 'onlineSavingsMsg'
Learn More

Ethylcyclohexane, 99+%, Thermo Scientific Chemicals

Product Code. 10550751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10550751 25 mL Glass bottle 25mL
10437110 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10550751 Supplier Thermo Scientific Acros Supplier No. 118310250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1678-91-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00001523
InChI Key IIEWJVIFRVWJOD-UHFFFAOYSA-N
Synonym cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100
PubChem CID 15504
IUPAC Name ethylcyclohexane
SMILES CCC1CCCCC1

Spécification

Melting Point -111°C
Color Colorless
Density 0.7800g/mL
Boiling Point 130°C to 132°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H11C2H5
Refractive Index 1.4311 to 1.4331
Quantity 25 mL
Beilstein 05, 35
Specific Gravity 0.78
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,diethyl ether,acetone,,benzene,carbon tetrachloride,lignoin
Formula Weight 112.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethylcyclohexane, 99+%
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Ethylcyclohexane
Mention d'avertissement
  • Danger
Catégorie de danger
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Mention de danger
  • H304-May be fatal if swallowed and enters airways.
  • H225-Highly flammable liquid and vapour.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.