missing translation for 'onlineSavingsMsg'
Learn More

ethyle1-(2-furylmethyl)-4,5-dioxopyrrolidine-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10495692
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10495692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10495692 Supplier Thermo Scientific Maybridge Supplier No. KM02778DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 142774-43-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H13NO5
MDL Number MFCD00085072
Quantity 1 g
Synonym ethyl 1-2-furylmethyl-4,5-dioxopyrrolidine-3-carboxylate, ethyl 1-furan-2-ylmethyl-4,5-dioxopyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylicacid, 1-2-furanylmethyl-4,5-dioxo-, ethyl ester, acmc-1c782, ethyl 1-2-furylmethyl-4,5-dioxoazolidine-3-carboxylate, ethyl 1-furan-2-yl methyl-4,5-dioxopyrrolidine-3-carboxylate, 3-pyrrolidinecarboxylicacid,1-2-furanylmethyl-4,5-dioxo-,ethyl ester, 1-furan-2-ylmethyl-4-hydroxy-5-oxo-2,5-dihydro-1h-pyrrole-3-carboxylic acid ethyl ester, 3-6-fluoro-1,1-dioxido-2-piperidin-1-ylcarbonyl-4h-1,4-benzothiazin-4-yl benzonitrile
InChI Key YGYRGGBYAUAMKP-UHFFFAOYSA-N
SMILES CCOC(=O)C1CN(C(=O)C1=O)CC2=CC=CO2
IUPAC Name ethyl 1-(furan-2-ylmethyl)-4,5-dioxopyrrolidine-3-carboxylate
Molecular Weight (g/mol) 251.238
PubChem CID 2778688
Formula Weight 251.24
Percent Purity 97%
Chemical Name or Material ethyle1-(2-furylmethyl)-4, 5-dioxopyrrolidine-3-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.