missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(2-methyl-1,3-thiazol-4-yl)acetate, 95%, Thermo Scientific™

Product Code. 10789783
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10789783 1 g 1g
10788435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10789783 Supplier Thermo Scientific Maybridge Supplier No. CD09887DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 37128-24-8
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00052519
InChI Key DWSMURKWFWKPBT-UHFFFAOYSA-N
Synonym ethyl 2-2-methyl-1,3-thiazol-4-yl acetate, 2-methyl-thiazol-4-yl-acetic acid ethyl ester, 2-methylthiazole-4-acetic acid ethyl ester, ethyl 2-methyl-1,3-thiazol-4-yl acetate, ethyl 2-2-methylthiazol-4-yl acetate, 4-thiazoleacetic acid,2-methyl-, ethyl ester, enamine_005349, ethyl=2-methylthiazol-4-ylacetate, 2-methylthiazol-4-yl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-methyl-, ethyl ester
PubChem CID 854815
IUPAC Name ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(=N1)C

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 185.25
Percent Purity 95%
Chemical Name or Material Ethyl 2-(2-methyl-1, 3-thiazol-4-yl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.