missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(5-bromo-2-thienyl)-2-oxoacetate, 97%, Thermo Scientific™

Product Code. 10408182
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10408182

Brand: Thermo Scientific Maybridge BTB09133DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22098-10-8
Molecular Formula C8H7BrO3S
Molecular Weight (g/mol) 263.105
MDL Number MFCD00085056
InChI Key PMBGHMBDWGNJJE-UHFFFAOYSA-N
Synonym ethyl 2-5-bromothiophen-2-yl-2-oxoacetate, ethyl 5-bromothien-2-yl glyoxylate, ethyl 2-5-bromo-2-thienyl-2-oxoacetate, ethyl 5-bromothiophene-2-glyoxylate, ethyl 5-bromo-2-thienyl glyoxylate, ethyl 2-5-bromothien-2-yl glyoxalate, ethyl 5-bromothiophen-2-yl oxo acetate, 2-bromo-5-ethoxy oxo acetyl thiophene, 5-bromothiophen-2-yl oxoacetic acid ethyl ester
PubChem CID 2736376
IUPAC Name ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CC=C(S1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 263.11
Percent Purity 97%
Chemical Name or Material Ethyl 2-(5-bromo-2-thienyl)-2-oxoacetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.