missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(5-bromo-2-thienyl)-2-oxoacetate, 97%, Thermo Scientific™

Product Code. 10512092
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10512092 10 g 10g
10408182 1 g 1g
10011274 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10512092 Supplier Thermo Scientific Maybridge Supplier No. BTB09133EA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 22098-10-8
Molecular Formula C8H7BrO3S
Molecular Weight (g/mol) 263.105
MDL Number MFCD00085056
InChI Key PMBGHMBDWGNJJE-UHFFFAOYSA-N
Synonym ethyl 2-5-bromothiophen-2-yl-2-oxoacetate, ethyl 5-bromothien-2-yl glyoxylate, ethyl 2-5-bromo-2-thienyl-2-oxoacetate, ethyl 5-bromothiophene-2-glyoxylate, ethyl 5-bromo-2-thienyl glyoxylate, ethyl 2-5-bromothien-2-yl glyoxalate, ethyl 5-bromothiophen-2-yl oxo acetate, 2-bromo-5-ethoxy oxo acetyl thiophene, 5-bromothiophen-2-yl oxoacetic acid ethyl ester
PubChem CID 2736376
IUPAC Name ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CC=C(S1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 263.11
Percent Purity 97%
Chemical Name or Material Ethyl 2-(5-bromo-2-thienyl)-2-oxoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.