missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate, 97%, Thermo Scientific™

Product Code. 10497222
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10497222 10 g 10g
10683393 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10497222 Supplier Thermo Scientific Maybridge Supplier No. CC00243EA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 306934-99-6
Molecular Formula C13H12BrNO2S
Molecular Weight (g/mol) 326.208
MDL Number MFCD00435056
InChI Key SEWFWRCESBYGFS-UHFFFAOYSA-N
PubChem CID 727636
IUPAC Name ethyl 2-amino-4-(4-bromophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=C(C=C2)Br)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Synonym ethyl 2-amino-4-4-bromophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylate, 2-amino-4-4-bromo-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-bromophenyl-thiophene-3-carboxylic acid ethyl ester, ethyl2-amino-4-4-bromophenyl-3-thiophenecarboxylate, ethyl2-amino-4-4-bromophenyl thiophene-3-carboxylate, 2-amino-4-4-bromophenyl-3-ethoxycarbonyl thiophene, ethyl 2-amino-4-4-bromophenyl-3-thiophene carboxylate
Formula Weight 326.21
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(4-bromophenyl)-3-thiophenecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.