missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-4-(4-chlorophenyl)-3-thiophenecarboxylate, 97%, Thermo Scientific™

Product Code. 10421283
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421283 10 g Amber Glass Bottle 10g
10137252 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421283 Supplier Thermo Scientific Maybridge Supplier No. MO07018EA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 65234-09-5
Molecular Formula C13H12ClNO2S
Molecular Weight (g/mol) 281.75
MDL Number MFCD01038372
InChI Key YHJWJJKFFNCDCK-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-chlorophenyl thiophene-3-carboxylate, 2-amino-4-4-chloro-phenyl-thiophene-3-carboxylicacidethylester, 2-amino-4-4-chlorophenyl thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-chloro-phenyl-thiophene-3-carboxylic acid ethyl ester, us8889672, apogee a, 2-amino-4-4-chlorophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-chlorophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-chlorophenyl-3-thiophenecarboxylate #, ethyl 2-amino-4-4-chlorophenyl-thiophene-3-carboxylate, 3-thiophenecarboxylicacid,2-amino-4-4-chlorophenyl-,ethyl ester
PubChem CID 616598
IUPAC Name ethyl 2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 281.76
Percent Purity 97%
Chemical Name or Material ethyle2-amino-4-(4-chlorophenyl)-3-thiophenecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.