missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-amino-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10131022
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10131022 1 g Amber Glass Bottle 1g
10029111 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10131022 Supplier Thermo Scientific Maybridge Supplier No. CD10070DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 81633-29-6
Molecular Formula C8H11N3O2
Molecular Weight (g/mol) 181.20
MDL Number MFCD00052622
InChI Key YBFVMJRSZCVJJP-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyrimidine-5-carboxylic acid ethyl ester, 2-amino-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, acmc-209plx, maybridge1_003176, ethyl2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-5-ethoxycarbonyl-4-methylpyrimidine, ethyl 2-amino-4-methyl-pyrimidine-5-carboxylate, ethyl 2-amino-4-methylpyrimidine-5-carboxylate, 2-amino-4-methyl-5-pyrimidinecarboxylic acid, ethyl ester, 5-pyrimidinecarboxylicacid, 2-amino-4-methyl-, ethyl ester
PubChem CID 459813
IUPAC Name ethyl 2-amino-4-methylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N)N=C1C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.19
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylpyrimidine-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.