missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-bromo-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10242742
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10242742 1 g Amber Glass Bottle 1g
10703443 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10242742 Supplier Thermo Scientific Maybridge Supplier No. MO00750DA
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 22900-83-0
Molecular Formula C7H8BrNO2S
Molecular Weight (g/mol) 250.11
MDL Number MFCD03791227
InChI Key CFBIOWPDDZPIDP-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4-methylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-4-methyl-, ethyl ester, 2-bromo-4-methyl-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4-methyl-5-thiazolecarboxylate, 2-bromo-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-bromo-4-methylthiazole-5-carboxylic acid ethyl ester, maybridge3_004543, ksc207i5t, ethyl2-bromo-4-methyl-1,3-thiazole-5-carboxylate, ethylbromomethylthiazolecarboxylate
PubChem CID 2824057
IUPAC Name ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Br)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 250.12
Percent Purity 97%
Chemical Name or Material ethyle2-bromo-4-methyl-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.