missing translation for 'onlineSavingsMsg'
Learn More

ethyle3-amino-4-(isopropylsulfonyl)-5-(methylthio)thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10099081
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10099081 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10099081 Supplier Thermo Scientific Maybridge Supplier No. KM03883DA
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175202-72-9
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H17NO4S3
MDL Number MFCD00085052
Quantity 1 g
Synonym ethyl 3-amino-4-isopropylsulfonyl-5-methylthio thiophene-2-carboxylate, ethyl 3-amino-5-methylsulfanyl-4-propane-2-sulfonyl thiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-amino-4-1-methylethyl sulfonyl-5-methylthio-, ethyl ester, maybridge1_004975, ethyl 3-amino-4-isopropylsulfonyl-5-methylthio thiophene-2carboxylate, ethyl 3-amino-4-isopropylsulphonyl-5-methylthio thiophene-2-carboxylate, 2-thiophenecarboxylicacid,3-amino-4-1-methylethyl sulfonyl-5-methylthio-,ethyl ester, 3-amino-5-methylthio-4-propan-2-ylsulfonyl-2-thiophenecarboxylic acid ethyl ester, ethyl 3-azanyl-5-methylsulfanyl-4-propan-2-ylsulfonyl-thiophene-2-carboxylate, ethyl3-amino-4-isopropylsulfonyl-5-methylthio thiophene-2-carboxylate
InChI Key RUTILLJYJHVEPU-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(N)C(=C(SC)S1)S(=O)(=O)C(C)C
IUPAC Name ethyl 3-amino-5-(methylsulfanyl)-4-(propane-2-sulfonyl)thiophene-2-carboxylate
Molecular Weight (g/mol) 323.44
PubChem CID 2778831
Formula Weight 323.46
Percent Purity 97%
Chemical Name or Material Ethyl 3-amino-4-(isopropylsulfonyl)-5-(methylthio)thiophene-2-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.