missing translation for 'onlineSavingsMsg'
Learn More

ethyle3-amino-4-(methylamino)benzoate, 97%, Thermo Scientific™

Product Code. 10364052
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10364052 10 g Amber Glass Bottle 10g
10354052 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10364052 Supplier Thermo Scientific Maybridge Supplier No. MO00109EA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 66315-23-9
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.23
MDL Number MFCD04110945
InChI Key ZPJHHBPBCFRECW-UHFFFAOYSA-N
PubChem CID 2824047
IUPAC Name ethyl 3-amino-4-(methylamino)benzoate
SMILES CCOC(=O)C1=CC(N)=C(NC)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym ethyl 3-amino-4-methylamino benzoate, 3-amino-4-methylamino benzoic acid ethyl ester, ethyl 4-methylamino-3-aminobenzoate, ethyl 3-azanyl-4-methylamino benzoate, ethyl3-amino-4-methylamino benzoate, 3-amino-4-methylaminobenzoic acid ethyl ester, benzoic acid,3-amino-4-methylamino-, ethyl ester, benzoic acid, 3-amino-4-methylamino-, ethyl ester
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material ethyle3-amino-4-(methylamino)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.