missing translation for 'onlineSavingsMsg'
Learn More

ethyle4,6-dichloro-2-(methylthio)quinoline-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10602263
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10602263 1 g Amber Glass Bottle 1g
10561122 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10602263 Supplier Thermo Scientific Maybridge Supplier No. KM08739DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 227958-96-5
Molecular Formula C13H11Cl2NO2S
Molecular Weight (g/mol) 316.20
MDL Number MFCD00276616
InChI Key NRGNGBKXHHRCBD-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloro-2-methylthio quinoline-3-carboxylate, ethyl 4,6-dichloro-2-methylsulfanyl quinoline-3-carboxylate, 3-quinolinecarboxylicacid, 4,6-dichloro-2-methylthio-, ethyl ester, pubchem6084, ethyl 4,6-dichloro-2-methylsulphanyl quinoline, 4,6-dichloro-3-ethoxycarbonyl-2-methylthio quinoline, 4,6-dichloro-2-methylthio-3-quinolinecarboxylic acid ethyl ester, ethyl 4,6-bis chloranyl-2-methylsulfanyl-quinoline-3-carboxylate
PubChem CID 2736409
SMILES CCOC(=O)C1=C(SC)N=C2C=CC(Cl)=CC2=C1Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name ethyl 4,6-dichloro-2-methylsulfanylquinoline-3-carboxylate
Formula Weight 316.21
Percent Purity 97%
Chemical Name or Material ethyle4, 6-dichloro-2-(methylthio)quinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.