missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10713243
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10713243 1 g 1g
10100642 10 g 10g
10524984 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10713243 Supplier Thermo Scientific Maybridge Supplier No. CC23010DA
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5408-07-1
Molecular Formula C9H12BrNO2
Molecular Weight (g/mol) 246.10
MDL Number MFCD02647051
InChI Key POLBZQUAKJYNIF-UHFFFAOYSA-N
Synonym 1h-pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl-, ethyl ester, 4-bromo-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 4-bromo-3,5-dimethylpyrrole-2-carboxylate, 4-bromo-3,5-dimethyl-2-ethoxycarbonyl-1h-pyrrole, ethyl 4-bromo-3,5-dimethyl-1h-pyrrole-2-carboxylat, pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl-, ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-bromo-3,5-dimethyl,ethyl ester
PubChem CID 138486
IUPAC Name ethyl 4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C(=C(N1)C)Br)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 246.1
Percent Purity 97%
Chemical Name or Material Ethyl 4-bromo-3, 5-dimethyl-1H-pyrrole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.