missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-formyl-2,5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate, 97%, Thermo Scientific™

Product Code. 10170892
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10170892 1 g 1g
10618925 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10170892 Supplier Thermo Scientific Maybridge Supplier No. SP00028DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175276-52-5
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD00204225
InChI Key BGDYIZGTPVRWAM-UHFFFAOYSA-N
Synonym ethyl 4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylate, 4-formyl-2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxylic acid ethyl ester
PubChem CID 2798484
IUPAC Name ethyl 4-formyl-2,5-dimethyl-1-phenylpyrrole-3-carboxylate
SMILES CCOC(=O)C1=C(N(C(=C1C=O)C)C2=CC=CC=C2)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 271.31
Percent Purity 97%
Chemical Name or Material Ethyl 4-formyl-2, 5-dimethyl-1-phenyl-1H-pyrrole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.