missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10793423
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10793423 1 g 1g
10314192 250 mg 250mg
10514984 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793423 Supplier Thermo Scientific Maybridge Supplier No. CC23004DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2199-64-6
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00030384
InChI Key CLJUICOFPKFFGJ-UHFFFAOYSA-N
Synonym 4-formyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 3,5-dimethyl-4-formylpyrrole-2-carboxylate, ethyl 4-formyl-3,5-dimethylpyrrole-2-carboxylate, 2,4-dimethyl-3-formyl-5-carbethoxypyrrole, 3,5-dimethyl-2-ethoxycarbonyl-4-formyl-1h-pyrrole, 3,5-dimethyl-4-formylpyrrole-2-carboxylic acid ethyl ester, ethyl4-formyl-3,5-dimethyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, 4-formyl-3,5-dimethyl-, ethyl ester, 1h-pyrrole-2-carboxylicacid,4-formyl-3,5-dimethyl-,ethyl ester, 3,5-dimethyl-4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester
PubChem CID 137486
IUPAC Name ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C)C(C=O)=C(C)N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 195.22
Percent Purity 97%
Chemical Name or Material ethyle4-formyl-3, 5-dimethyl-1H-pyrrole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.