missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10713853
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10713853 10 g Amber Glass Bottle 10g
10631893 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713853 Supplier Thermo Scientific Maybridge Supplier No. SPB02157EA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 53715-64-3
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD00141953
InChI Key FYPLITQTMHJFKK-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-phenylthiazole-5-carboxylate, maybridge1_008302, ethyl4-methyl-2-phenyl-1,3-thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-5-thiazolecarboxylate, ethyl 4-methyl-2-phenyl thiazole-5-carboxylate, ethyl 4-methyl-2-phenyl-thiazole-5-carboxylate, 5-ethoxycarbonyl-4-methyl-2-phenyl-1,3-thiazole, 2-phenyl-4-methylthiazole-5-carboxylic acid ethyl ester, 4-methyl-2-phenyl-5-thiazolecarboxylic acid ethyl ester, 4-methyl-2-phenyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 2743970
IUPAC Name ethyl 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)C2=CC=CC=C2)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyl-2-phenyl-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.