missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10190552
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10190552 10 g 10g
10181272 1 g 1g
10031144 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190552 Supplier Thermo Scientific Maybridge Supplier No. RH00119EA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 138907-68-3
Molecular Formula C12H12FN3O2
Molecular Weight (g/mol) 249.25
MDL Number MFCD00173917
InChI Key RPPPCKSHIYWAPW-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
PubChem CID 688689
IUPAC Name ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 249.24
Percent Purity 97%
Chemical Name or Material ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.