missing translation for 'onlineSavingsMsg'
Learn More

Ethylene chlorophosphite, 97%

Product Code. 11490663
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490663 500 g 500g
11470663 25 g 25g
11480663 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490663 Supplier Thermo Scientific Alfa Aesar Supplier No. A12724.36
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 822-39-9 | C2H4ClO2P | 126.476 g/mol

Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.

Notes
Store in cool place. Moisture sensitive. Incompatible with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-39-9
Molecular Formula C2H4ClO2P
Molecular Weight (g/mol) 126.476
MDL Number MFCD00014511
InChI Key OLSFRDLMFAOSIA-UHFFFAOYSA-N
PubChem CID 69973
IUPAC Name 2-chloro-1,3,2-dioxaphospholane
SMILES C1COP(O1)Cl

Specifications

Density 1.415
Boiling Point 46°C to 47°C (15 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.491
Quantity 500 g
UN Number UN3265
Beilstein 506179
Sensitivity Moisture sensitive
Synonym ethylene chlorophosphite, 1,3,2-dioxaphospholane, 2-chloro, ethylene cyclo-chlorophosphite, acmc-209pnx, 1,3,2-dioxaphospholane,2-chloro, 2-chloro-1,3,2-dioxaphospholane, ethylene phosphorochloridite 2-chloro-1,3,2-dioxap, inchi=1/c2h4clo2p/c3-6-4-1-2-5-6/h1-2h
Formula Weight 126.47
Percent Purity 97%
Chemical Name or Material Ethylene chlorophosphite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.