missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene-d4 Thiourea, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Imidazolidinethione-4,5-d4 |
| CAS | 352431-28-8 |
| Molecular Formula | C3 2H4 H2 N2 S |
| Quantity | 0.05 g |
| Synonym | 2-Imidazolidinethione-4,4,5,5-d4 (9 CI), Ethylene thiourea D4 |
| InChI Formula | InChI=1 S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])NC(=S)NC1([2 H])[2 H] |
| IUPAC Name | 4,4,5,5-tetradeuterioimidazolidine-2-thione |
| Molecular Weight (g/mol) | 106.18 |
| Formula Weight | 106.0503 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction