missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Dicysteine Diester (ECD) Dihydrochloride, TRC

Product Code. 30011816
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011816 25 mg 25mg
30012664 10 mg 10mg
30005051 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011816 Supplier Toronto Research Chemicals Supplier No. TRCC14344025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14344-58-2
Molecular Formula C12H24N2O4S2 . 2(HCl)
Molecular Weight (g/mol) 397.38
Synonym N,N'-1,2-Ethanediylbis[L-cysteine]-1,1'-diethyl ester hydrochloride, (2R,2'R)-Diethyl 2,2'-(ethane-1,2-diylbis(azanediyl))bis(3-mercaptopropanoate)
IUPAC Name ethyl (2R)-2-[2-[[(2R)-1-ethoxy-1-oxo-3-sulfanylpropan-2-yl]amino]ethylamino]-3-sulfanylpropanoate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)[C@H](CS)NCCN[C@@H](CS)C(=O)OCC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H24N2O4S2.2ClH/c1-3-17-11(15)9(7-19)13-5-6-14-10(8-20)12(16)18-4-2;;/h9-10,13-14,19-20H,3-8H2,1-2H3;2*1H/t9-,10-;;/m0../s1
Chemical Name or Material Ethylene Dicysteine Diester (ECD) Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.