missing translation for 'onlineSavingsMsg'
Learn More

Ethylene dimethacrylate, 98%, stabilized

Product Code. p-129031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
This item is not returnable. View return policy

Product Code. 10607201

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-90-5 | C10H14O4 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 98.0°C to 100.0°C (5.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (H2C=C(CH3)COOCH2-)2
Refractive Index 1.4530 to 1.4550
Quantity 250 mL
Specific Gravity 1.05
Solubility Information Solubility in water: soluble.
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
Stabilizer 90 to 110ppm MEHQ
Viscosity 3.1 mPa.s (20°C)
Formula Weight 198.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene dimethacrylate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.