missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 95%, pure

Product Code. 10107470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10107470 1 kg Glass Bottle 1kg
11924631 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107470 Supplier Thermo Scientific Acros Supplier No. 410010010
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Quantity 1 kg
Flash Point 116°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Viscous Liquid
Refractive Index 1.4300 to 1.4330
Assay Percent Range 95%
Specific Gravity 1.113
Linear Formula HOCH2CH2OH
Viscosity 21 mPa.s (20°C)
Fieser 01,375; 05,296; 09,217; 15,156
Boiling Point 196.0°C to 198.0°C
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Formula Weight 62.06
Percent Purity 95%
Grade Pure
Chemical Name or Material Ethylene glycol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.