missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, BAKER ANALYZED™, J.T. Baker™

Product Code. 10579112 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10579112 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10579112 Supplier JT Baker Supplier No. 7037.1000
  • ONLINE EXCLUSIVE PRICE
    82.00 €
    78.27 € / 1 L
    Save 3.73 €
    4% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point -13°C
CAS 107-21-1
Color Max. 10 (APHA)
Acidity max. 0.01%
Density 1.115 g/cm³ (20°C)
Quantity 1 L
Flash Point 116°C
Packaging Glass Bottle
Assay Percent Range min. 99.0%
Molecular Formula C2H6O2
MDL Number MFCD00002885
Water max. 0.2%
Boiling Point 196°C to 198°C (1013 hPa)
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.