missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol-Bis-(Beta-Aminoethylether)-N,N,N',N'-Tetraacetic Acid Crystalline MP Biomedicals

Product Code. 11471062 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471062 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471062 Supplier MP Biomedicals Supplier No. 0219517480
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-42-5
Quantity 100 g
Physical Form Crystalline
Molecular Formula C14H24N2O10
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 380.35
PubChem CID 6207
ChEBI CHEBI:30740
Formula Weight 380.4
Chemical Name or Material Ethylene Glycol-Bis-(Beta-Aminoethylether)-N,N,N',N'-Tetraacetic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.