missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Butyl Ether, ≥99.0%, Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™

Product Code. 15672760
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15672760 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15672760 Supplier Honeywell Riedel-de Haën Supplier No. 2563661L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 111-76-2
Density 0.902g/mL at 25°C
Explosion Limit 0.106
Flash Point 67°C (152.6°F)
Ignition Point 473°F
Molecular Formula C6H14O2
Linear Formula CH3(CH2)3OCH2CH2OH
MDL Number MFCD00002884
Quantity 1 L
Beilstein 1732511
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
SMILES CCCCOCCO
IUPAC Name 2-butoxyethan-1-ol
Molecular Weight (g/mol) 118.18
PubChem CID 8133
Vapor Density 4.1 (vs air)
ChEBI CHEBI:63921
Formula Weight 118.17g/mol
Vapor Pressure 1mmHg (20°C), 10mmHg (81°C)
Percent Purity ≥99.0%
Grade Sprectrophotometric
Chemical Name or Material Ethylene Glycol Butyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.