missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol

Product Code. 11349658
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
4 kg
Unit Size:
1kg
250g
4kg
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349658 4 kg 4kg
11319658 50 g 50g
11329658 250 g 250g
11339658 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11349658 Supplier Thermo Scientific Alfa Aesar Supplier No. 044151.A6
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Crosslinking monomer | CAS: 97-90-5 | C10H14O4 | 198.22 g/mol

Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Color Colorless to Yellow
Density 1.051 g/mL
Boiling Point 83°C to 85°C (1 mmHg)
Flash Point >110°C (230°F)
Linear Formula H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
Refractive Index 1.454
Quantity 4 kg
Beilstein 1776663
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 198.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene glycol dimethacrylate, Stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.