missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT

Produktkod. 10396550 Handla allt< titel> Produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Förpackningsstorlek:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
10396550 500 mL Glass Bottle 500mL
10696682 1 L Glass Bottle 1 L
10606692 2.5 L Glass Bottle 2.5 L
10247752 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10396550 missing translation for 'mfr' Thermo Scientific missing translation for 'supplierNo' 118455000
  • ONLINE EXCLUSIVE PRICE
    75.30 €
    69.20 € / 500mL
    Save 6.10 €
    8% Off
Visa alla kampanjer
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylene glycol dimethyl ether
pH 6 to 7.5 (1:9)
Melting Point -69.0°C
Name Note Stabilized with BHT
CAS 128-37-0
Density 0.8670g/mL
Quantity 500 mL
Flash Point −6°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3785 to 1.3805
Assay Percent Range 99+%
Specific Gravity 0.867
Linear Formula CH3OCH2CH2OCH3
Viscosity 1.1 mPa.s (20°C)
Water 0.1% max. (K.F.)
Beilstein 01, 467
Fieser 01,267; 02,143; 16,229
Merck Index 15, 3245
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
Boiling Point 84.0°C to 86.0°C
Solubility Information Solubility in water: miscible. Other solubilities: solulbe in hydrocarbon solvents, soluble in ethanol, ethyl ether, acetone, carbon, tetrachloride, benzene, dichloroethane
Formula Weight 90.12
Percent Purity ≥99%
Grade Extra Pure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol dimethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.