missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, High Purity, Low Iron and Low Chloride, Greenfield

Product Code. 30286169
Change view
Click to view available options
Quantity:
4x1 Gallon
5 Gallon
Unit Size:
15.14 L
18.93 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286169 4x1 Gallon 15.14 L
30286179 5 Gallon 18.93 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30286169 Supplier Pharmco Supplier No. 371000000CSGL
  • ONLINE EXCLUSIVE PRICE
    890.82 €
    877.17 € / 15.14 L
    Save 13.65 €
    1% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 107-21-1

Specifications

Quantity 4x1 Gallon
Packaging 4x1 Gallon Case
Boiling Point 197.3°C
Chemical Name or Material Ethylene Glycol, High Purity, Low Iron and Low Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.