missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell Riedel-de Haen™
BIOPHARMA

Product Code. 16605733
Change view
Click to view available options
30 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16605733 30 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605733 Supplier Honeywell Riedel-de Haën Supplier No. 0375030KG
  • ONLINE EXCLUSIVE PRICE
    1023.87 €
    883.62 € / 30kg
    Save 140.25 €
    13% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point -12°C
CAS 107-21-1
Color Colorless
CAS Max % 1
Density 1.11 g/cm3 (20°C)
Quantity 30 kg
Flash Point 111°C
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Beilstein 505945
Synonym 1,2-Ethanediol
Boiling Point 197°C
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Grade Pharmacopeia
Chemical Name or Material Ethylene glycol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.