missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Product Code. 16655733 Shop All Honeywell Research Chemicals Products
Click to view available options
:
25 kg
This item is not returnable. View return policy

Product Code. 16655733

Brand: Honeywell Research Chemicals 10246625KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point -12°C
CAS 107-21-1
Color Colorless
CAS Max % 1
Density 1.11 g/cm3 (20°C)
Boiling Point 197°C
Quantity 25 kg
Flash Point 111°C
Molecular Formula C2H6O2
Linear Formula HOCH2CH2OH
Beilstein 505945
Synonym 1,2-Ethanediol
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Grade Reagent
Chemical Name or Material Ethylene glycol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.