missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Solstice

Product Code. 15690960 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
20 L
225kg
30kg
5 L
500mL
Pack of 4
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15690960 4 x 2.5 L Poly Bottle Pack of 4
15600970 500 mL Poly Bottle 500mL
15630960 1 L Poly Bottle 1 L
15640960 2.5 L Poly Bottle 2.5 L
15620970 6 x 500 mL Poly Bottle Pack of 6
15610970 5 L Poly Bottle 5 L
15650960 20 L Stainless Steel Can 20 L
15680960 30 kg Stainless Steel Can 30kg
15670960 225 kg Metal Drum 225kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15690960 Supplier Fluka Supplier No. 1024664X2.5L
  • ONLINE EXCLUSIVE PRICE
    675.00 €
    552.77 € / Pack of 4
    Save 122.23 €
    18% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C (literature)
CAS Max % 100.0000%
Density 1.113 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 111°C
Packaging Poly Bottle
Linear Formula HOCH2CH2OH
Beilstein 505945
Boiling Point 197°C
Formula Weight 62.07
Percent Purity >99%
Grade Reagent
Chemical Name or Material Ethylene glycol, Reagent Grade, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.