missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Terephthalate Cyclic Heptamer, TRC

Product Code. 30012424
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012424 2.5 mg 2.50mg
30005675 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012424 Supplier Toronto Research Chemicals Supplier No. TRCE9181752.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29668-12-0
Molecular Formula C70 H56 O28
Molecular Weight (g/mol) 1345.177
Synonym 3,6,13,16,23,26,33,36,43,46,53,56,63,66-Tetradecaoxaoctacyclo[66.2.2.28,11.218,21.228,31.238,41.248,51.258,61]tetraoctaconta-8,10,18,20,28,30,38,40,48,50,58,60,68,70,71,73,75,77,79,81,83-heneicosaene-2,7,12,17,22,27,32,37,42,47,52,57,62,67-tetradecone, Terephthalic Acid, Heptamol. Cyclic Ethylene Ester, Ethylene Glycol, Heptamol. Cyclic Terephthalate
SMILES O=C1OCCOC(=O)C2=CC=C(C=C2)C(=O)OCCOC(=O)C3=CC=C(C=C3)C(=O)OCCOC(=O)C4=CC=C(C=C4)C(=O)OCCOC(=O)C5=CC=C(C=C5)C(=O)OCCOC(=O)C6=CC=C(C=C6)C(=O)OCCOC(=O)C7=CC=C(C=C7)C(=O)OCCOC(=O)C8=CC=C1C=C8

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C70H56O28/c71-57-43-1-2-44(4-3-43)58(72)86-31-32-88-60(74)46-9-11-48(12-10-46)62(76)90-35-36-92-64(78)50-17-19-52(20-18-50)66(80)94-39-40-96-68(82)54-25-27-56(28-26-54)70(84)98-42-41-97-69(83)55-23-21-53(22-24-55)67(81)95-38-37-93-65(79)51-15-13-49(14-16-51)63(77)91-34-33-89-61(75)47-7-5-45(6-8-47)59(73)87-30-29-85-57/h1-28H,29-42H2
Formula Weight 1344.296
Percent Purity >95%
Chemical Name or Material Ethylene Terephthalate Cyclic Heptamer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.