missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Terephthalate Cyclic Hexamer, TRC

Product Code. 30383156
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383156 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383156 Supplier Toronto Research Chemicals Supplier No. TRCE9181801MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Ethylene Terephthalate Cyclic Hexamer
CAS 29644-29-9
Molecular Formula C60 H48 O24
Quantity 1 mg
Synonym 3,6,13,16,23,26,33,36,43,46,53,56-Dodecaoxaheptacyclo[56.2.2.28,11.218,21.228,31.238,41.248,51]doheptaconta-8,10,18,20,28,30,38,40,48,50,58,60,61,63,65,67,69,71-octadecaene-2,7,12,17,22,27,32,37,42,47,52,57-dodecone (9CI), Ethylene glycol, hexamol. cyclic
InChI Formula InChI=1S/C60H48O24/c61-49-37-1-2-38(4-3-37)50(62)74-27-28-76-52(64)40-9-11-42(12-10-40)54(66)78-31-32-80-56(68)44-17-19-46(20-18-44)58(70)82-35-36-84-60(72)48-23-21-47(22-24-48)59(71)83-34-33-81-57(69)45-15-13-43(14-16-45)55(67)79-30-29-77-53(65)41-7-5-39
SMILES O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCOC(=O)c4ccc(cc4)C(=O)OCCOC(=O)c5ccc(cc5)C(=O)OCCOC(=O)c6ccc(cc6)C(=O)OCCOC(=O)c7ccc1cc7
Molecular Weight (g/mol) 1153.009
Formula Weight 1152.254
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.