missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(isothiouronium bromide), 98%, Thermo Scientific™

Product Code. 11316996
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316996 5 g 5g
11326996 25 g 25g
11336996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316996 Supplier Thermo Scientific Alfa Aesar Supplier No. L06784.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6943-65-3
Molecular Formula C4H12Br2N4S2
Molecular Weight (g/mol) 340.096
MDL Number MFCD00035055
InChI Key VABTXEYLYWGJRB-UHFFFAOYSA-N
Synonym ethylenediisothiouronium dibromide, ethylene diisothiouronium dibromide, ethylenediisothiourea dihydrobromide, ethylene diisothiourea dihydrobromide, pseudourea, 2,2-ethylenedithiodi-, dihydrobromide, 2,2'-ethylenebisisothiouronium dibromide, carbamimidothioic acid, 1,2-ethanediyl ester, dihydrobromide, ethylenebis isothiouronium bromide, 2,2'-ethylene-bis-2-thiopseudourea , dihydrobromide, pseudourea, 2,2'-ethylenebis 2-thio-, dihydrobromide
PubChem CID 81370
IUPAC Name 2-carbamimidoylsulfanylethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)SC(=N)N.Br.Br

Specifications

Melting Point 224°C to 226°C
Quantity 5 g
Beilstein 3914616
Sensitivity Hygroscopic
Formula Weight 340.09
Percent Purity 98%
Chemical Name or Material Ethylenebis(isothiouronium bromide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.